missing translation for 'onlineSavingsMsg'
Learn More

Protamine Sulfate from Salmon sperm, TCI America™

Catalog No. P06751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06751G 1 g
P06755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06751G Supplier TCI America Supplier No. P06751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53597-25-4
MDL Number MFCD00132091
Synonym Salmine Sulfate

Specifications

Physical Form Powder/Crystals
Color White
pH 4.0 to 7.0 (1% H2O soln.)
Quantity 1 g
Chemical Name or Material Protamine Sulfate from Salmon sperm
RTECSNumber VO9470000
TSCA No