missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Prostratin, 99+%

Supplier:  Thermo Scientific Chemicals J60971MCR

 View more versions of this product

Catalog No. AAJ60971MCR


Only null left

Description

Description

12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1ug per mouse

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1 mg
-30°C to -10°C
99+%
C22H30O6
216°C to 219°C
Powder
C[C@@H]1CC2(OC(C)=O)C([C@@H]3C=C(CO)C[C@@]4(O)[C@@H](C=C(C)C4=O)[C@@]13O)C2(C)C
390.48
White
12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1μg per mouse
60857-08-1
390.47
Soluble in DMSO at 30mg/ml
BOJKFRKNLSCGHY-KKFGZBDXSA-N
(1R,2S,6R,10S,15R)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-13-yl acetate

Chemical Identifiers

60857-08-1
C22H30O6
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only