missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Prostratin, 99+%
Description
12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1ug per mouse
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 mg |
| Description | White |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | 12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1μg per mouse |
| Formula Weight | 390.47 |
| Melting Point | 216°C to 219°C |
| Solubility | Soluble in DMSO at 30mg/ml |
| Form | Powder |
| InChI Key | BOJKFRKNLSCGHY-KKFGZBDXSA-N |
| SMILES | C[C@@H]1CC2(OC(C)=O)C([C@@H]3C=C(CO)C[C@@]4(O)[C@@H](C=C(C)C4=O)[C@@]13O)C2(C)C |
| Show More |
Safety and Handling
| Recommended Storage | Store at -20°C |
RUO – Research Use Only