missing translation for 'onlineSavingsMsg'
Learn More

Propyzamide 96.0+%, TCI America™

Catalog No. P23745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23745G 5 g
P23741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23745G Supplier TCI America Supplier No. P23745G
Only null left

Chemical Identifiers

CAS 23950-58-5
Molecular Formula C12H11Cl2NO
Molecular Weight (g/mol) 256.126
MDL Number MFCD00055346
InChI Key PHNUZKMIPFFYSO-UHFFFAOYSA-N
Synonym propyzamide, pronamide, 3,5-dichloro-n-1,1-dimethylpropynyl benzamide, 3,5-dichloro-n-2-methylbut-3-yn-2-yl benzamide, kerb, pronamid, clanex, 3,5-dichloro-n-1,1-dimethyl-2-propynyl benzamide, kerb 50w, caswell no. 306a
PubChem CID 32154
ChEBI CHEBI:34935
IUPAC Name 3,5-dichloro-N-(2-methylbut-3-yn-2-yl)benzamide
SMILES CC(C)(C#C)NC(=O)C1=CC(=CC(=C1)Cl)Cl

Specifications

Melting Point 157°C
Color White
Quantity 5 g
UN Number 3077
Formula Weight 256.13
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Propyzamide
RTECSNumber CV3460000
TSCA No
Recommended Storage Refrigerator