missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyzamide 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 157°C |
| Color | White |
| Quantity | 1 g |
| UN Number | 3077 |
| Formula Weight | 256.13 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Propyzamide |
Safety and Handling
| RTECSNumber | CV3460000 |
| TSCA | No |
| Recommended Storage | Refrigerator |