missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylparaben Sodium, NF, 94-102%, Spectrum™ Chemical
P1457, 35285-69-9, C10H11NaO3
$239.80 - $22301.97
Chemical Identifiers
| CAS | 35285-69-9 |
|---|---|
| Molecular Formula | C10H11NaO3 |
| Molecular Weight (g/mol) | 202.19 |
| InChI Key | IXMINYBUNCWGER-UHFFFAOYSA-M |
| IUPAC Name | sodium 4-(propoxycarbonyl)benzen-1-olate |
| SMILES | [Na+].CCCOC(=O)C1=CC=C([O-])C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605929
|
Spectrum Chemical Mfg Cor
P145725GM |
25 g | Poly Bottle |
Each for $239.80
|
|
||||
|
18605925
|
Spectrum Chemical Mfg Cor
P1457500GM |
500 g | Poly Bottle |
Each for $1,012.31
|
|
||||
|
18605926
|
Spectrum Chemical Mfg Cor
P145725KG |
2.5 kg | Poly Pail |
Each for $2,707.23
|
|
||||
|
18605927
|
Spectrum Chemical Mfg Cor
P145712KG |
12 kg | Poly Pail |
Each for $7,730.69
|
|
||||
|
18605928
|
Spectrum Chemical Mfg Cor
P145750KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Propylparaben Sodium, NF is used as an antimicrobial preservative. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 35285-69-9 | |
| 202.19 | |
| sodium 4-(propoxycarbonyl)benzen-1-olate |
| C10H11NaO3 | |
| IXMINYBUNCWGER-UHFFFAOYSA-M | |
| [Na+].CCCOC(=O)C1=CC=C([O-])C=C1 |
Specifications
| 35285-69-9 | |
| 9.5 to 10.5 | |
| C10H11NaO3 | |
| IXMINYBUNCWGER-UHFFFAOYSA-M | |
| sodium 4-(propoxycarbonyl)benzen-1-olate | |
| 94 to 102% |
| 1 | |
| Poly Bottle | |
| 25 g | |
| [Na+].CCCOC(=O)C1=CC=C([O-])C=C1 | |
| 202.19 | |
| NF |