missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylparaben Sodium, NF, 94-102%, Spectrum™ Chemical
P1457, 35285-69-9, C10H11NaO3
Supplier: Spectrum Chemical Mfg Cor P145712KG
Description
Spectrum™ Chemical Propylparaben Sodium, NF is used as an antimicrobial preservative. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 35285-69-9 | |
| 9.5 to 10.5 | |
| C10H11NaO3 | |
| IXMINYBUNCWGER-UHFFFAOYSA-M | |
| sodium 4-(propoxycarbonyl)benzen-1-olate | |
| 94 to 102% |
| 1 | |
| Poly Pail | |
| 12 kg | |
| [Na+].CCCOC(=O)C1=CC=C([O-])C=C1 | |
| 202.19 | |
| NF |
Chemical Identifiers
| 35285-69-9 | |
| 202.19 | |
| sodium 4-(propoxycarbonyl)benzen-1-olate |
| C10H11NaO3 | |
| IXMINYBUNCWGER-UHFFFAOYSA-M | |
| [Na+].CCCOC(=O)C1=CC=C([O-])C=C1 |