missing translation for 'onlineSavingsMsg'
Learn More

Medisca Propylparaben NF

Catalog No. 14185531
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185531 500 g
14185480 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14185531 Supplier Medisca Supplier No. 155108*

Propylparaben NF is a small, colorless crystals or white powder. Alternative name(s): propyl hydroxybenzoate, Propyl paraben, Propyl parahydroxybenzoate

Propylparaben NF is a small, colorless crystals or white powder.

Alternative name(s): propyl hydroxybenzoate, Propyl paraben, Propyl parahydroxybenzoate

Specifications

Chemical Name or Material Propylparaben NF
Packaging Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 94-13-3
Type Drug Standard
Molecular Weight (g/mol) 180.202
Quantity 500 g
Percent Purity 98 to 102 %
Grade NF