missing translation for 'onlineSavingsMsg'
Learn More

Propylene Sulfide (stabilized with Butyl Mercaptan) 98.0+%, TCI America™

Catalog No. p03215ml
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03215ML 5 mL
P032125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03215ML Supplier TCI America Supplier No. P03215ML
Only null left

Chemical Identifiers

CAS 1072-43-1
Molecular Formula C3H6S
Molecular Weight (g/mol) 74.14
MDL Number MFCD00005160
InChI Key MBNVSWHUJDDZRH-UHFFFAOYNA-N
Synonym propylene sulfide, thiirane, methyl, thiirane, 2-methyl, propene sulfide, 1,2-epithiopropane, propylene sulphide, propylene episulfide, 2-methylthiacyclopropane, propane, 1,2-epithio, methylthiirane
PubChem CID 14072
IUPAC Name 2-methylthiirane
SMILES CC1CS1

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 mL
UN Number 1992
Formula Weight 74.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propylene Sulfide (stabilized with Butyl Mercaptan)
RTECSNumber TZ2625000
TSCA Yes