missing translation for 'onlineSavingsMsg'
Learn More

Propylene Phenoxytol, Reagent, Spectrum™ Chemical

Catalog No. 18605922
Change view
Click to view available options
Quantity:
25 mL
125 mL
4 L
20 L
200 L
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605922 200 L Poly Bottle
18605918 25 mL Poly Bottle
18605923 125 mL Poly Bottle
18605920 4 L Poly Bottle
18605921 20 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18605922 Supplier Spectrum Chemical Mfg Cor Supplier No. PR132200LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PR132, 770-35-4, C9H12O2

Spectrum™ Chemical Propylene Phenoxytol, Reagent

Chemical Identifiers

CAS 770-35-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00016861
InChI Key IBLKWZIFZMJLFL-UHFFFAOYNA-N
IUPAC Name 1-phenoxypropan-2-ol
SMILES CC(O)COC1=CC=CC=C1

Specifications

CAS Max % 1
Boiling Point 242 to 244°C
Packaging Poly Bottle
Quantity 200 L
Percent Purity 93%
Grade Reagent
Physical Form Liquid