missing translation for 'onlineSavingsMsg'
Learn More

Medisca Propylene Glycol USP

Catalog No. 14185671
Change view
Click to view available options
Quantity:
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185671 20 L
14185651 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14185671 Supplier Medisca Supplier No. 051007
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Propylene Glycol USP is a clear, colorless, viscous liquid. It is practically odorless. Absorbs moisture when exposed to moist air.

Propylene Glycol USP is a clear, colorless, viscous liquid. It is practically odorless. Absorbs moisture when exposed to moist air.

Alternative name(s): 1,2-Propanediol, Monopropylene glycol, Mono propylene glycol, Propylene, Propane-1,2-diol, Propan-1,2-diol, methylethyl glycol

Specifications

Chemical Name or Material Propylene Glycol USP
Packaging Pail, 6 gallon, White, HDPE, Round
CAS 57-55-6
Type Drug Standard
Molecular Weight (g/mol) 76.09
Quantity 20 L
Percent Purity ≥99.5 %
Grade USP