missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol Monostearate 83.0+%, TCI America™

Supplier:  TCI America P216325G

 View more versions of this product

Catalog No. P216325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Propylene Glycol Monostearate
44°C
C21H42O3
Hydroxypropyl Stearate, Stearic Acid Hydroxypropyl Ester
≥83.0% (GC)
1323-39-3
White
25 g
342.56
Crystalline Powder

Chemical Identifiers

1323-39-3
C21H42O3
Hydroxypropyl Stearate, Stearic Acid Hydroxypropyl Ester
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0772&(2)-2523

RTECSNumber : WI4550000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions