missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Glycol Monostearate 83.0+%, TCI America™
$63.52 - $238.72
Chemical Identifiers
| CAS | 1323-39-3 |
|---|---|
| Molecular Formula | C21H42O3 |
| Synonym | Hydroxypropyl Stearate, Stearic Acid Hydroxypropyl Ester |
Chemical Identifiers
| 1323-39-3 | |
| C21H42O3 | |
| Hydroxypropyl Stearate, Stearic Acid Hydroxypropyl Ester |
Specifications
| 1323-39-3 | |
| White | |
| 25 g | |
| 342.56 | |
| Crystalline Powder |
| 44°C | |
| C21H42O3 | |
| Hydroxypropyl Stearate, Stearic Acid Hydroxypropyl Ester | |
| ≥83.0% (GC) | |
| Propylene Glycol Monostearate |
Safety and Handling
EINECSNumber : (2)-0772&(2)-2523
RTECSNumber : WI4550000
TSCA : Yes