missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol, 85% (v/v), Ricca Chemical

Catalog No. R6583900500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6583900500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R6583900500 Supplier Ricca Chemical Company Supplier No. R6583900500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-55-6 , 7732-18-5
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Color Colorless
Physical Form Liquid
CAS Min % 84.0, 14.26
CAS Max % 87.0, 14.85
Quantity 500 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 85% (v/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Propylene Glycol
ShelfLife 24 months