missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Carbonate 98.0+%, TCI America™
Supplier: TCI America P052525G
Specifications
| Propylene Carbonate | |
| -49°C | |
| 242°C | |
| MFCD00005385,MFCD00798264,MFCD00798265 | |
| propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate | |
| CC1COC(=O)O1 | |
| 102.09 | |
| 102.09 | |
| Liquid |
| 108-32-7 | |
| Yellow | |
| C4H6O3 | |
| 25 g | |
| RUOJZAUFBMNUDX-UHFFFAOYNA-N | |
| 4-methyl-1,3-dioxolan-2-one | |
| 7924 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 108-32-7 | |
| 102.09 | |
| RUOJZAUFBMNUDX-UHFFFAOYNA-N | |
| 7924 | |
| CC1COC(=O)O1 |
| C4H6O3 | |
| MFCD00005385,MFCD00798264,MFCD00798265 | |
| propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate | |
| 4-methyl-1,3-dioxolan-2-one |
Safety and Handling
EINECSNumber : (5)-0524&(7)-0737
RTECSNumber : FF9650000
TSCA : Yes