missing translation for 'onlineSavingsMsg'
Learn More

Propylene Carbonate 98.0+%, TCI America™

Catalog No. p-7134659
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
P052525G 25 g
P0525500G 500 g
2 options

Catalog No. P052525G

Supplier: TCI America P052525G

Only null left

Chemical Identifiers

CAS 108-32-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00005385,MFCD00798264,MFCD00798265
InChI Key RUOJZAUFBMNUDX-UHFFFAOYNA-N
Synonym propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
PubChem CID 7924
IUPAC Name 4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Melting Point -49°C
Color Yellow
Boiling Point 242°C
Quantity 25 g
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propylene Carbonate
EINECSNumber (5)-0524&(7)-0737
RTECSNumber FF9650000
TSCA Yes