Learn More
Propylene carbonate, 99.5%
Propylene carbonate, 99.5%, Quantity: 2.5L, Packaging: Glass bottle, Boiling Point: 240.0 deg.C to 243.0 deg.C, Undesignated, Melting Point: -49.0 deg.C, Molecular Weight: 102.09, Percent Purity: 99.5%, Assay Percent Range: 99.4% min. (GC), CAS: 108-32-7, Density: 1.2040g/mL | CAS: 108-32-7 | C4H6O3 | 102.09 g/mol
Supplier: Thermo Scientific Chemicals 131560100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
| Propylene carbonate | |
| -49.0°C | |
| 240.0°C to 243.0°C | |
| Authentic | |
| Plastic drum | |
| 1.4200 to 1.4220 | |
| 10 L | |
| propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate | |
| RUOJZAUFBMNUDX-UHFFFAOYNA-N | |
| 4-methyl-1,3-dioxolan-2-one | |
| 7924 | |
| 102.09 | |
| Liquid |
| 108-32-7 | |
| 1.2040g/mL | |
| 116°C | |
| 99.4% min. (GC) | |
| C4H6O3 | |
| MFCD00005385,MFCD00798264,MFCD00798265 | |
| 1.204 | |
| Solubility in water: 200g/L (25°C). Other solubilities: miscible with acetone,benzene,chloroform,dmf,,ether,ethyl acetate and ethyl alcohol,slightly soluble in carbon tetrachloride,soluble in most organic solvents | |
| CC1COC(=O)O1 | |
| 102.09 | |
| 2.8 mPa.s (20°C) | |
| 99.5% |
Chemical Identifiers
| 108-32-7 | |
| 102.09 | |
| RUOJZAUFBMNUDX-UHFFFAOYNA-N | |
| 7924 | |
| CC1COC(=O)O1 |
| C4H6O3 | |
| MFCD00005385,MFCD00798264,MFCD00798265 | |
| propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate | |
| 4-methyl-1,3-dioxolan-2-one |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-572-1
RUO – Research Use Only