missing translation for 'onlineSavingsMsg'
Learn More

Propylene carbonate, 99.5%

Catalog No. p-3737273
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131565000 500 mL Glass bottle
AC131560010 1 L Glass bottle
AC131560025 2.5 L Glass bottle
AC131560100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC131565000 Supplier Thermo Scientific Chemicals Supplier No. 131565000
Only null left

Propylene carbonate, 99.5%, Quantity: 2.5L, Packaging: Glass bottle, Boiling Point: 240.0 deg.C to 243.0 deg.C, Undesignated, Melting Point: -49.0 deg.C, Molecular Weight: 102.09, Percent Purity: 99.5%, Assay Percent Range: 99.4% min. (GC), CAS: 108-32-7, Density: 1.2040g/mL | CAS: 108-32-7 | C4H6O3 | 102.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 108-32-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00005385,MFCD00798264,MFCD00798265
InChI Key RUOJZAUFBMNUDX-UHFFFAOYNA-N
Synonym propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
PubChem CID 7924
IUPAC Name 4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Melting Point -49.0°C
Density 1.2040g/mL
Boiling Point 240.0°C to 243.0°C
Flash Point 116°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Refractive Index 1.4200 to 1.4220
Quantity 500 mL
Specific Gravity 1.204
Solubility Information Solubility in water: 200g/L (25°C). Other solubilities: miscible with acetone,benzene,chloroform,dmf,,ether,ethyl acetate and ethyl alcohol,slightly soluble in carbon tetrachloride,soluble in most organic solvents
Viscosity 2.8 mPa.s (20°C)
Formula Weight 102.09
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Propylene carbonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-572-1

RUO – Research Use Only