missing translation for 'onlineSavingsMsg'
Learn More

Propylamine Hydroiodide 97.0+%, TCI America™

Catalog No. p-7135457
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22121G 1 g
P22125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22121G Supplier TCI America Supplier No. P22121G
Only null left

Chemical Identifiers

CAS 14488-45-0
Molecular Formula C3H10IN
Molecular Weight (g/mol) 187.024
InChI Key GIAPQOZCVIEHNY-UHFFFAOYSA-N
Synonym Propylammonium Iodide
PubChem CID 91972164
IUPAC Name propan-1-amine;hydroiodide
SMILES CCCN.I

Specifications

Color White
Quantity 1 g
Formula Weight 187.02
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Propylamine Hydroiodide