accessibility menu, dialog, popup

UserName

CAS RN 14488-45-0

IUPAC Name: propan-1-amine hydroiodide
Synonyms: propylamine hydroiodide
Molecular Weight (g/mol): 187.02
Molecular Formula: C3H10IN
InChi Key: GIAPQOZCVIEHNY-UHFFFAOYSA-N
SMILES: I.CCCN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results