missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylamine Hydrobromide 97.0+%, TCI America™
Supplier: TCI America P25025G
Specifications
| Propylamine Hydrobromide | |
| 182°C | |
| C3H10BrN | |
| Propylammonium Bromide | |
| CCCN.Br | |
| 140.024 | |
| 140.02 | |
| Crystalline Powder |
| 4905-83-3 | |
| White | |
| 5 g | |
| MVYQJCPZZBFMLF-UHFFFAOYSA-N | |
| propan-1-amine;hydrobromide | |
| 12203661 | |
| ≥97.0% (N,T) |
Chemical Identifiers
| 4905-83-3 | |
| 140.024 | |
| Propylammonium Bromide | |
| propan-1-amine;hydrobromide |
| C3H10BrN | |
| MVYQJCPZZBFMLF-UHFFFAOYSA-N | |
| 12203661 | |
| CCCN.Br |
Safety and Handling
TSCA : No