missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylamine Hydrobromide 97.0+%, TCI America™
$50.33 - $134.21
Chemical Identifiers
| CAS | 4905-83-3 |
|---|---|
| Molecular Formula | C3H10BrN |
| Molecular Weight (g/mol) | 140.024 |
| InChI Key | MVYQJCPZZBFMLF-UHFFFAOYSA-N |
| Synonym | Propylammonium Bromide |
| PubChem CID | 12203661 |
| IUPAC Name | propan-1-amine;hydrobromide |
| SMILES | CCCN.Br |
Chemical Identifiers
| 4905-83-3 | |
| 140.024 | |
| Propylammonium Bromide | |
| propan-1-amine;hydrobromide |
| C3H10BrN | |
| MVYQJCPZZBFMLF-UHFFFAOYSA-N | |
| 12203661 | |
| CCCN.Br |
Specifications
| 4905-83-3 | |
| White | |
| 1 g | |
| MVYQJCPZZBFMLF-UHFFFAOYSA-N | |
| propan-1-amine;hydrobromide | |
| 12203661 | |
| ≥97.0% (N,T) | |
| Propylamine Hydrobromide |
| 182°C | |
| C3H10BrN | |
| Propylammonium Bromide | |
| CCCN.Br | |
| 140.024 | |
| 140.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : No