missing translation for 'onlineSavingsMsg'
Learn More

Propyl Sulfide 98.0+%, TCI America™

Catalog No. P053225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P053225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P053225ML Supplier TCI America Supplier No. P053225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-47-7
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00009379
InChI Key ZERULLAPCVRMCO-UHFFFAOYSA-N
Synonym propyl sulfide, dipropyl sulfide, di-n-propyl sulfide, propane, 1,1'-thiobis, dipropyl thioether, propyl monosulfide, dipropyl sulphide, n-propyl sulfide, 4-thiaheptane, 1,1'-thiobispropane
PubChem CID 8118
IUPAC Name 1-(propylsulfanyl)propane
SMILES CCCSCCC

Specifications

Melting Point 0°C
Color Colorless
Boiling Point 142°C
Quantity 25 mL
UN Number 1993
Formula Weight 118.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Sulfide