missing translation for 'onlineSavingsMsg'
Learn More

Propyl Methanesulfonate 98.0+%, TCI America™

Catalog No. P18445G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18445G 5 g
P184425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18445G Supplier TCI America Supplier No. P18445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1912-31-8
Molecular Formula C4H10O3S
Molecular Weight (g/mol) 138.181
MDL Number MFCD00007561
InChI Key DKORSYDQYFVQNS-UHFFFAOYSA-N
Synonym Methanesulfonic Acid Propyl Ester
PubChem CID 74705
IUPAC Name propyl methanesulfonate
SMILES CCCOS(=O)(=O)C

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 138.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Methanesulfonate
RTECSNumber PB2680000
TSCA No