accessibility menu, dialog, popup

UserName

H3COSCH3OO

CAS RN 1912-31-8

IUPAC Name: propyl methanesulfonate
Synonyms: propyl methanesulfonate
Molecular Weight (g/mol): 138.18
Molecular Formula: C4H10O3S
InChi Key: DKORSYDQYFVQNS-UHFFFAOYSA-N
SMILES: CCCOS(C)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results