missing translation for 'onlineSavingsMsg'
Learn More

Propyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I03885ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03885ML 5 mL
I038825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03885ML Supplier TCI America Supplier No. I03885ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-30-8
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD00004823
InChI Key KKASGUHLXWAKEZ-UHFFFAOYSA-N
Synonym propyl isothiocyanate, propylisothiocyanate, propane, 1-isothiocyanato, n-propyl isothiocyanate, isothiocyanic acid propyl ester, isothiocyanic acid, propyl ester, isothiocyanic acid n-propyl ester, 1-propyl isothiocyanate, propanisothiocyanate, 1-isothiocyanatopropan
PubChem CID 69403
IUPAC Name 1-isothiocyanatopropane
SMILES CCCN=C=S

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 mL
UN Number 2924
Formula Weight 101.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Isothiocyanate