missing translation for 'onlineSavingsMsg'
Learn More

Propyl gallate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012785
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012785 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012785 Supplier Merck Emd Millipore Supplier No. PHR11181G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Propyl gallate is a synthetic phenolic antioxidant (SPA) utilized in food additives to inhibit or regulate the lipid oxidation. SPAs are cost-effective, largely available and highly efficient in comparison to their natural counterparts.

Specifications

CAS 121-79-9
Flash Point 187°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula 3,4,5-(HO)3C6H2 CO2 CH2 CH2 CH3
MDL Number MFCD00002196
Melting Point 146°C to 149°C
Synonym 3,4,5-Trihydroxybenzoic acid propyl ester
Type Certified Reference Material
Quantity 1 g
Formula Weight 212.20
Grade Certified Reference Material
Show More Show Less