Learn More
Propyl Gallate, FCC, 98-102%, Spectrum™ Chemical
P1461, 121-79-9, C10H12O5
Supplier:  Spectrum Chemical Mfg Cor P146125KG
 View more versions of this product
Catalog No. 18605898
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Propyl Gallate, FCC is used to protect oils and fats in products from oxidation. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
121-79-9
2.5 kg
CCCOC(=O)C1=CC(O)=C(O)C(O)=C1
212.2
FCC
146-150°C
ZTHYODDOHIVTJV-UHFFFAOYSA-N
propyl 3,4,5-trihydroxybenzoate
98 to 102%
Chemical Identifiers
121-79-9
ZTHYODDOHIVTJV-UHFFFAOYSA-N
CCCOC(=O)C1=CC(O)=C(O)C(O)=C1
212.2
propyl 3,4,5-trihydroxybenzoate
Product Suggestions
Product Suggestions