missing translation for 'onlineSavingsMsg'
Learn More

Propyl Gallate, FCC, 98-102%, Spectrum™ Chemical

Catalog No. p-7158105
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605896 100 g Poly Bottle
18605897 500 g Poly Bottle
18605898 2.5 kg -
18605899 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605896 Supplier Spectrum Chemical Mfg Cor Supplier No. P1461100GM
Only null left

P1461, 121-79-9, C10H12O5

Spectrum™ Chemical Propyl Gallate, FCC is used to protect oils and fats in products from oxidation. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 121-79-9
Molecular Weight (g/mol) 212.2
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N
IUPAC Name propyl 3,4,5-trihydroxybenzoate
SMILES CCCOC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Melting Range 146-150°C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade FCC