missing translation for 'onlineSavingsMsg'
Learn More

Propyl Formate 95.0+%, TCI America™

Catalog No. F005825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F005825ML 25 mL
F0058500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F005825ML Supplier TCI America Supplier No. F005825ML

Chemical Identifiers

CAS 110-74-7
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00003295
InChI Key KFNNIILCVOLYIR-UHFFFAOYSA-N
Synonym formic acid, propyl ester, n-propyl formate, propyl methanoate, formiate de propyle, formic acid propyl ester, propylester kyseliny mravenci, unii-wo1arv6gtw, formiate de propyle french, fema no. 2943, wo1arv6gtw
PubChem CID 8073
IUPAC Name propyl formate
SMILES CCCOC=O

Specifications

Color Yellow
Boiling Point 81°C
Quantity 25 mL
UN Number 1281
Formula Weight 88.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Formate
EINECSNumber (2)-0684
RTECSNumber LR0175000
TSCA Yes