accessibility menu, dialog, popup

UserName

H3COO

CAS RN 110-74-7

IUPAC Name: propyl formate
Synonyms: propyl formate
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: KFNNIILCVOLYIR-UHFFFAOYSA-N
SMILES: CCCOC=O
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results