missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl Cyanoacetate 99.0+%, TCI America™
Supplier: TCI America C1016500ML
Specifications
| Propyl Cyanoacetate | |
| Yellow | |
| C6H9NO2 | |
| 500 mL | |
| NLFIMXLLXGTDME-UHFFFAOYSA-N | |
| propyl 2-cyanoacetate | |
| 84444 | |
| ≥99.0% (GC) |
| 14447-15-5 | |
| 216°C | |
| MFCD00015784 | |
| Cyanoacetic Acid Propyl Ester | |
| CCCOC(=O)CC#N | |
| 127.14 | |
| 127.14 | |
| Liquid |
Chemical Identifiers
| 14447-15-5 | |
| 127.14 | |
| NLFIMXLLXGTDME-UHFFFAOYSA-N | |
| 84444 | |
| CCCOC(=O)CC#N |
| C6H9NO2 | |
| MFCD00015784 | |
| Cyanoacetic Acid Propyl Ester | |
| propyl 2-cyanoacetate |
Safety and Handling
EINECSNumber : (2)-1549
TSCA : No