missing translation for 'onlineSavingsMsg'
Learn More

Propyl Cyanoacetate 99.0+%, TCI America™

Catalog No. p-7124744
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C101625ML 25 mL
C1016500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C101625ML Supplier TCI America Supplier No. C101625ML

Chemical Identifiers

CAS 14447-15-5
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.14
MDL Number MFCD00015784
InChI Key NLFIMXLLXGTDME-UHFFFAOYSA-N
Synonym Cyanoacetic Acid Propyl Ester
PubChem CID 84444
IUPAC Name propyl 2-cyanoacetate
SMILES CCCOC(=O)CC#N

Specifications

Color Yellow
Boiling Point 216°C
Quantity 25 mL
Formula Weight 127.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl Cyanoacetate
EINECSNumber (2)-1549
TSCA No