missing translation for 'onlineSavingsMsg'
Learn More

Propyl Benzoate 99.0+%, TCI America™

$52.19 - $158.44

Chemical Identifiers

CAS 2315-68-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00009370
InChI Key UDEWPOVQBGFNGE-UHFFFAOYSA-N
Synonym n-propyl benzoate, benzoic acid, propyl ester, benzoic acid n-propyl ester, propyl benzenecarboxylate, benzoic acid propyl ester, unii-vwk210b7ws, benzoate de propyle, vwk210b7ws, dsstox_cid_24878, dsstox_rid_80552
PubChem CID 16846
IUPAC Name propyl benzoate
SMILES CCCOC(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B007625ML
View Documents
TCI America
B007625ML
25 mL N/A
Only null left
 
B0076500ML
View Documents
TCI America
B0076500ML
500 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2315-68-6
164.204
UDEWPOVQBGFNGE-UHFFFAOYSA-N
16846
CCCOC(=O)C1=CC=CC=C1
C10H12O2
MFCD00009370
n-propyl benzoate, benzoic acid, propyl ester, benzoic acid n-propyl ester, propyl benzenecarboxylate, benzoic acid propyl ester, unii-vwk210b7ws, benzoate de propyle, vwk210b7ws, dsstox_cid_24878, dsstox_rid_80552
propyl benzoate

Specifications

2315-68-6
230°C
MFCD00009370
n-propyl benzoate, benzoic acid, propyl ester, benzoic acid n-propyl ester, propyl benzenecarboxylate, benzoic acid propyl ester, unii-vwk210b7ws, benzoate de propyle, vwk210b7ws, dsstox_cid_24878, dsstox_rid_80552
CCCOC(=O)C1=CC=CC=C1
164.204
164.20
Liquid
Colorless
C10H12O2
25 mL
UDEWPOVQBGFNGE-UHFFFAOYSA-N
propyl benzoate
16846
≥99.0% (GC)
Propyl Benzoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1356

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'