missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl Acetoacetate 98.0+%, TCI America™
$96.26 - $96.26
Chemical Identifiers
| CAS | 1779-60-8 |
|---|---|
| Molecular Formula | C7H12O3 |
| Molecular Weight (g/mol) | 144.17 |
| MDL Number | MFCD00059405 |
| InChI Key | DHGFMVMDBNLMKT-UHFFFAOYSA-N |
| Synonym | Acetoacetic Acid Propyl Ester |
| PubChem CID | 74507 |
| IUPAC Name | propyl 3-oxobutanoate |
| SMILES | CCCOC(=O)CC(C)=O |
Chemical Identifiers
| 1779-60-8 | |
| 144.17 | |
| DHGFMVMDBNLMKT-UHFFFAOYSA-N | |
| 74507 | |
| CCCOC(=O)CC(C)=O |
| C7H12O3 | |
| MFCD00059405 | |
| Acetoacetic Acid Propyl Ester | |
| propyl 3-oxobutanoate |
Specifications
| 1779-60-8 | |
| 194°C | |
| MFCD00059405 | |
| Acetoacetic Acid Propyl Ester | |
| CCCOC(=O)CC(C)=O | |
| 144.17 | |
| 144.17 | |
| Liquid |
| Colorless | |
| C7H12O3 | |
| 25 mL | |
| DHGFMVMDBNLMKT-UHFFFAOYSA-N | |
| propyl 3-oxobutanoate | |
| 74507 | |
| ≥98.0% (GC) | |
| Propyl Acetoacetate |
Safety and Handling
EINECSNumber : (2)-1475
TSCA : No