missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl Acetoacetate 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 194°C |
| Quantity | 25 mL |
| Formula Weight | 144.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Propyl Acetoacetate |
Safety and Handling
| EINECSNumber | (2)-1475 |
| TSCA | No |