missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl Acetate 98.0+%, TCI America™
Supplier: TCI America A0044500ML
Specifications
| Propyl Acetate | |
| Colorless | |
| C5H10O2 | |
| 500 mL | |
| n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester | |
| CCCOC(C)=O | |
| 102.13 | |
| CHEBI:40116 | |
| ≥98.0% (GC) |
| 109-60-4 | |
| 101°C | |
| MFCD00009372 | |
| 1276 | |
| YKYONYBAUNKHLG-UHFFFAOYSA-N | |
| propyl acetate | |
| 7997 | |
| 102.13 | |
| Liquid |
Chemical Identifiers
| 109-60-4 | |
| 102.13 | |
| YKYONYBAUNKHLG-UHFFFAOYSA-N | |
| 7997 | |
| propyl acetate |
| C5H10O2 | |
| MFCD00009372 | |
| n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester | |
| CHEBI:40116 | |
| CCCOC(C)=O |
Safety and Handling
EINECSNumber : (2)-0727
RTECSNumber : AJ3675000
TSCA : Yes