missing translation for 'onlineSavingsMsg'
Learn More

Propyl 2-Furancarboxylate 97.0+%, TCI America™

Catalog No. F015525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F015525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F015525ML Supplier TCI America Supplier No. F015525ML
Only null left

Chemical Identifiers

CAS 615-10-1
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00046108
InChI Key HSCVIIISAAEVQT-UHFFFAOYSA-N
Synonym 2-Furoic Acid Propyl Ester, 2-Furancarboxylic Acid Propyl Ester, Propyl 2-Furoate
PubChem CID 11982
IUPAC Name propyl furan-2-carboxylate
SMILES CCCOC(=O)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 211°C
Quantity 25 mL
Formula Weight 154.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Propyl 2-Furancarboxylate
EINECSNumber (5)-0046
RTECSNumber LV2022500
TSCA No