accessibility menu, dialog, popup

UserName

H3COOO

CAS RN 615-10-1

IUPAC Name: propyl furan-2-carboxylate
Synonyms: propyl furan-2-carboxylate
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: HSCVIIISAAEVQT-UHFFFAOYSA-N
SMILES: CCCOC(=O)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results