missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propranolol Hydrochloride 99.0+%, TCI America™
Supplier: TCI America P0995250G
Specifications
| Propranolol Hydrochloride | |
| 166°C | |
| C16H22ClNO2 | |
| 250 g | |
| ZMRUPTIKESYGQW-UHFFFAOYSA-N | |
| 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride | |
| 62882 | |
| 295.81 | |
| Crystalline Powder |
| 318-98-9 | |
| White | |
| MFCD00012558 | |
| propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal | |
| CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl | |
| 295.807 | |
| CHEBI:8500 | |
| ≥99.0% (HPLC,T) |
Chemical Identifiers
| 318-98-9 | |
| 295.807 | |
| ZMRUPTIKESYGQW-UHFFFAOYSA-N | |
| 62882 | |
| 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride |
| C16H22ClNO2 | |
| MFCD00012558 | |
| propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal | |
| CHEBI:8500 | |
| CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl |
Safety and Handling
RTECSNumber : UB7525000
TSCA : No