missing translation for 'onlineSavingsMsg'
Learn More

Propranolol Hydrochloride 99.0+%, TCI America™

Catalog No. p-7134891
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
P099525G 25 g
P0995250G 250 g
2 options

Catalog No. P099525G

Supplier: TCI America P099525G

Only null left

Chemical Identifiers

CAS 318-98-9
Molecular Formula C16H22ClNO2
Molecular Weight (g/mol) 295.807
MDL Number MFCD00012558
InChI Key ZMRUPTIKESYGQW-UHFFFAOYSA-N
Synonym propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal
PubChem CID 62882
ChEBI CHEBI:8500
IUPAC Name 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl

Specifications

Melting Point 166°C
Color White
Quantity 25 g
Formula Weight 295.81
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Propranolol Hydrochloride
RTECSNumber UB7525000
TSCA No