missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Propranolol hydrochloride, ≥98.5% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 318-98-9 |
| Physical Form | Neat |
| Linear Formula | C16H21 NO2 · HCl |
| MDL Number | MFCD00012558 |
| Synonym | (+/-)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride; DL-Propranolol hydrochloride |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 295.80 |
| Percent Purity | ≥98.5% (HPLC) |
| Grade | Analytical Standard |