missing translation for 'onlineSavingsMsg'
Learn More

Propiophenone, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015931
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015931 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015931 Supplier Merck Emd Millipore Supplier No. 610741ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 93-55-0
Vapor Pressure 1 mmHg (50°C)
Density 1.009 g/mL (at 25°C)
Flash Point 99°C - closed cup
Physical Form Neat
Linear Formula C6H5 COC2H5
MDL Number MFCD00009309
Melting Point 17°C to 19°C
Boiling Point 218°C
Synonym Ethyl phenyl ketone
Type Analytical Standard
Quantity 1 mL
Formula Weight 134.18
Percent Purity ≥97% (GC)
Grade Analytical Standard
Show More Show Less