missing translation for 'onlineSavingsMsg'
Learn More

Propionylcholine p-Toluenesulfonate 98.0+%, TCI America™

$192.94 - $2,321.87

Chemical Identifiers

CAS 1866-13-3
Molecular Formula C15H25NO5S
Molecular Weight (g/mol) 331.427
MDL Number MFCD00059975
InChI Key QEWHHEJZLZTUQT-UHFFFAOYSA-M
PubChem CID 44630436
IUPAC Name 4-methylbenzenesulfonate;trimethyl(2-propanoyloxyethyl)azanium
SMILES CCC(=O)OCC[N+](C)(C)C.CC1=CC=C(C=C1)S(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P05171G
View Documents
TCI America
P05171G
1 g N/A
Only null left
 
P051725G
View Documents
TCI America
P051725G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1866-13-3
331.427
QEWHHEJZLZTUQT-UHFFFAOYSA-M
4-methylbenzenesulfonate;trimethyl(2-propanoyloxyethyl)azanium
C15H25NO5S
MFCD00059975
44630436
CCC(=O)OCC[N+](C)(C)C.CC1=CC=C(C=C1)S(=O)(=O)[O-]

Specifications

1866-13-3
White-Yellow
MFCD00059975
QEWHHEJZLZTUQT-UHFFFAOYSA-M
4-methylbenzenesulfonate;trimethyl(2-propanoyloxyethyl)azanium
44630436
≥98.0% (HPLC,T)
Propionylcholine p-Toluenesulfonate
120°C
C15H25NO5S
1 g
CCC(=O)OCC[N+](C)(C)C.CC1=CC=C(C=C1)S(=O)(=O)[O-]
331.427
331.43
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents