missing translation for 'onlineSavingsMsg'
Learn More

Propionylcholine p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. P051725G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P051725G 25 g
P05171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P051725G Supplier TCI America Supplier No. P051725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1866-13-3
Molecular Formula C15H25NO5S
Molecular Weight (g/mol) 331.427
MDL Number MFCD00059975
InChI Key QEWHHEJZLZTUQT-UHFFFAOYSA-M
PubChem CID 44630436
IUPAC Name 4-methylbenzenesulfonate;trimethyl(2-propanoyloxyethyl)azanium
SMILES CCC(=O)OCC[N+](C)(C)C.CC1=CC=C(C=C1)S(=O)(=O)[O-]

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 25 g
Formula Weight 331.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Propionylcholine p-Toluenesulfonate