missing translation for 'onlineSavingsMsg'
Learn More

Propionyl-L-carnitine-(N-methyl-d3), ≥97.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111013472
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013472 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013472 Supplier Merck Emd Millipore Supplier No. 5294110MG

Specifications

CAS 203806-01-3
Linear Formula C10D3H16 NO4
MDL Number MFCD01073492
Synonym (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-(1-oxopropoxy)-1-propanaminium inner salt; L-Carnitine-(methyl-d3) propionyl ester; Propanoyl-L-carnitine-(N-methyl-d3)
Type Analytical Standard
Quantity 10 mg
Formula Weight 220.28
Percent Purity ≥97.0% (TLC)
Grade Analytical Standard