missing translation for 'onlineSavingsMsg'
Learn More

Propionitrile 99.0+%, TCI America™

Catalog No. P051425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P051425ML 25 mL
P0514100ML 100 mL
P0514500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P051425ML Supplier TCI America Supplier No. P051425ML

Chemical Identifiers

CAS 107-12-0
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00001948
InChI Key FVSKHRXBFJPNKK-UHFFFAOYSA-N
Synonym propionitrile, ethyl cyanide, propiononitrile, cyanoethane, propylnitrile, propionic nitrile, ether cyanatus, hydrocyanic ether, ethylkyanid, propannitril
PubChem CID 7854
ChEBI CHEBI:26307
IUPAC Name propanenitrile
SMILES CCC#N

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 mL
UN Number 2404
Formula Weight 55.08
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Propionitrile
EINECSNumber (2)-1509
RTECSNumber UF9625000
TSCA Yes