missing translation for 'onlineSavingsMsg'
Learn More

Propionitrile, 99%, extra pure

Catalog No. p-3737250
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC180891000 100 mL Glass bottle
AC180895000 500 mL Glass bottle
AC180890025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC180891000 Supplier Thermo Scientific Chemicals Supplier No. 180891000

CAS: 107-12-0 | C3H5N | 55.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-12-0
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00001948
InChI Key FVSKHRXBFJPNKK-UHFFFAOYSA-N
Synonym propionitrile, ethyl cyanide, propiononitrile, cyanoethane, propylnitrile, propionic nitrile, ether cyanatus, hydrocyanic ether, ethylkyanid, propannitril
PubChem CID 7854
ChEBI CHEBI:26307
IUPAC Name propanenitrile
SMILES CCC#N

Specifications

Melting Point -93.0°C
Density 0.7700g/mL
Boiling Point 97.0°C
Flash Point 6°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3650 to 1.3670
Linear Formula C2H5CN
Quantity 100 mL
Beilstein 02,245
Merck Index 15,7941
Specific Gravity 0.77
Solubility Information Solubility in water: 99.4g/L (20°C). Other solubilities: miscible with alcohol, ether, dmf, acetone,, benzene and dichloroethylen
Viscosity 0.44 mPa.s (20°C)
Formula Weight 55.08
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Propionitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Fatal in contact with skin.
Fatal if swallowed.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
EINECSNumber 203-464-4

RUO – Research Use Only