missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid, FCC, 99.5-100.5%, Spectrum™ Chemical

P1326, 79-09-4, C3H6O2

Supplier:  Spectrum Chemical Mfg Cor P13261LT

 View more versions of this product

Catalog No. 18605878


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Propionic Acid, FCC is used as a preservative and flavoring agent in packaged foods such as baked goods and cheese. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

79-09-4
Amber Glass Bottle
1 L
XBDQKXXYIPTUBI-UHFFFAOYSA-N
propanoic acid
99.5 to 100.5%
1
C3H6O2
0.993-0.997
CCC(O)=O
74.08
FCC

Chemical Identifiers

79-09-4
74.08
propanoic acid
C3H6O2
XBDQKXXYIPTUBI-UHFFFAOYSA-N
CCC(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions