missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid, FCC, 99.5-100.5%, Spectrum™ Chemical

Catalog No. p-7158556
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605877 500 mL Amber Glass Bottle
18605878 1 L Amber Glass Bottle
18605879 4 L Amber Glass Bottle
18605880 4 x 4 L Amber Glass Bottle
18605881 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18605877 Supplier Spectrum Chemical Mfg Cor Supplier No. P1326500ML
Only null left

P1326, 79-09-4, C3H6O2

Spectrum™ Chemical Propionic Acid, FCC is used as a preservative and flavoring agent in packaged foods such as baked goods and cheese. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 79-09-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
IUPAC Name propanoic acid
SMILES CCC(O)=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Specific Gravity 0.993-0.997
Percent Purity 99.5 to 100.5%
Grade FCC